accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 556-67-2

IUPAC Name: octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Synonyms: octamethylcyclotetrasiloxane oktamethylzyklotetrasiloxan union carbide 7207 octamethyl-cyclotetrasiloxane oktamethylcyklotetrasiloxan silicone SF 1173 nuc silicone VS 7207 octamethyltetrasiloxane omcts cyclic dimethylsiloxane tetramer
Molecular Weight (g/mol): 296.62
Molecular Formula: C8H24O4Si4
InChi Key: HMMGMWAXVFQUOA-UHFFFAOYSA-N
SMILES: C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results