accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiH3COSiCH3CH3OSiCH3CH3OSiCH3CH3OSiCH3H3COSiCH3H3COSiH3CH3COSiH3CH3COSiH3CH3CO

CAS RN 556-71-8

IUPAC Name: octadecamethylcyclononasiloxane
Synonyms: octadecamethylcyclononasiloxane
Molecular Weight (g/mol): 667.39
Molecular Formula: C18H54O9Si9
InChi Key: ISXOGOLHEGHGQF-UHFFFAOYSA-N
SMILES: C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)