accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 556-90-1

IUPAC Name: 2-amino-4,5-dihydro-1,3-thiazol-4-one
Synonyms: 4(5H)-thiazolone, 2-amino-
Molecular Weight (g/mol): 116.14
Molecular Formula: C3H4N2OS
InChi Key: HYMJHROUVPWYNQ-UHFFFAOYSA-N
SMILES: NC1=NC(=O)CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results