accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3Br

CAS RN 556-96-7

IUPAC Name: 1-bromo-3,5-dimethylbenzene
Synonyms: benzene, 1-bromo-3,5-dimethyl-
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: LMFRTSBQRLSJHC-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results