accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHNOO

CAS RN 5570-19-4

IUPAC Name: (2-nitrophenyl)boronic acid
Synonyms: 2-nitrophenylboronic acid
Molecular Weight (g/mol): 166.93
Molecular Formula: C6H6BNO4
InChi Key: SFUIGUOONHIVLG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results