accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOBrNOO

CAS RN 55715-03-2

IUPAC Name: 4-(bromomethyl)-3-nitrobenzoate
Synonyms: 4-(bromomethyl)-3-nitrobenzoate
Molecular Weight (g/mol): 259.04
Molecular Formula: C8H5BrNO4
InChi Key: QMAHVAFURJBOFV-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(CBr)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results