accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 55727-23-6

IUPAC Name: 3-(1,3-dithian-2-ylidene)pentane-2,4-dione
Synonyms: 3-(1,3-dithian-2-ylidene)pentane-2,4-dione
Molecular Weight (g/mol): 216.31
Molecular Formula: C9H12O2S2
InChi Key: PGPIRMKKHQOWIK-UHFFFAOYSA-N
SMILES: CC(=O)C(C(C)=O)=C1SCCCS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)