accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3H3CH3COHPOHOO

CAS RN 5574-97-0

IUPAC Name: tetrabutylazanium dihydrogen phosphate
Synonyms: tetrabutylammonium ion dihydrogen phosphate
Molecular Weight (g/mol): 339.46
Molecular Formula: C16H38NO4P
InChi Key: ARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILES: OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (3)
  • (15)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (6)
  • (1)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (15)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)