accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSiOCH3OCH3OCH3

CAS RN 5575-48-4

IUPAC Name: decyltrimethoxysilane
Synonyms: decyltrimethoxysilane
Molecular Weight (g/mol): 262.47
Molecular Formula: C13H30O3Si
InChi Key: KQAHMVLQCSALSX-UHFFFAOYSA-N
SMILES: CCCCCCCCCC[Si](OC)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results