accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOOONOO

CAS RN 55750-62-4

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3-(2,5-dioxopyrrol-1-yl)propanoate
Molecular Weight (g/mol): 266.21
Molecular Formula: C11H10N2O6
InChi Key: JKHVDAUOODACDU-UHFFFAOYSA-N
SMILES: O=C(CCN1C(=O)C=CC1=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)