accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5580-34-7

CH3NH2OH3C

CAS RN 5580-34-7

IUPAC Name: 2,3-dimethylbenzamide
Synonyms: 2,3-dimethylbenzamide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: IZAYISYTIWLBNB-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(N)=O)=C1C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)