accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

INHN

CAS RN 55919-82-9

IUPAC Name: 5-iodo-1H-indazole
Synonyms: 5-iodo-1H-indazole
Molecular Weight (g/mol): 244.04
Molecular Formula: C7H5IN2
InChi Key: CGCHCLICSHIAAM-UHFFFAOYSA-N
SMILES: IC1=CC=C2NN=CC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results