accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

A(a)

CAS RN 55963-78-5

IUPAC Name: sodium 1-methoxy-4-[(1E)-prop-1-en-1-yl]benzene sulfite
Synonyms: sodium anethole sulfite
Molecular Weight (g/mol): 251.25
Molecular Formula: C10H12NaO4S
InChi Key: JKJBFNAERWARKW-CZEFNJPISA-L
SMILES: [Na+].[O-]S([O-])=O.COC1=CC=C(\C=C\C)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results