accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NClClN

CAS RN 56-05-3

IUPAC Name: 4,6-dichloropyrimidin-2-amine
Synonyms: 4,6-dichloropyrimidin-2-amine
Molecular Weight (g/mol): 163.99
Molecular Formula: C4H3Cl2N3
InChi Key: JPZOAVGMSDSWSW-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=CC(Cl)=N1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)