accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56-34-8

IUPAC Name: tetraethylazanium chloride
Synonyms: tetraethylammonium chloride tetraethyl ammonium chloride
Molecular Weight (g/mol): 165.71
Molecular Formula: C8H20ClN
InChi Key: YMBCJWGVCUEGHA-UHFFFAOYSA-M
SMILES: [Cl-].CC[N+](CC)(CC)CC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results