accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ON(S)HO(R)(S)N(R)CH2

CAS RN 56-54-2

IUPAC Name: (S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol
Synonyms: quinidine quin-release nelson brand of quinidine sulfate apo-quinidine cardioquin (9S)-6'-methoxycinchonan-9-ol apotex brand of quinidine sulfate robins brand of quinidine sulfate biquin durules quinidine (product)
Molecular Weight (g/mol): 324.42
Molecular Formula: C20H24N2O2
InChi Key: LOUPRKONTZGTKE-LHHVKLHASA-N
SMILES: COC1=CC=C2N=CC=C([C@H](O)[C@H]3C[C@@H]4CCN3C[C@@H]4C=C)C2=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)