accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56-69-9

NHONH2OHHO

CAS RN 56-69-9

IUPAC Name: 2-amino-3-(5-hydroxy-1H-indol-3-yl)propanoic acid
Synonyms: hydroxytryptophan 5-hydroxy-DL-tryptophan DL-5-htp DL-5-hydroxytryptophan 5-hydroxytryptophan DL-form
Molecular Weight (g/mol): 220.23
Molecular Formula: C11H12N2O3
InChi Key: LDCYZAJDBXYCGN-UHFFFAOYNA-N
SMILES: NC(CC1=CNC2=CC=C(O)C=C12)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)