CAS RN 56-73-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-Glucose 6-Phosphate Solution (1M in H2O), TRC
CAS: 56-73-5 Molecular Formula: C6H13O9P Molecular Weight (g/mol): 260.14 Synonym: D-Glucose 6-Phosphate,Glucose 6-Phosphate,Robison Ester SMILES: O[C@@H]1[C@@H](O)[C@H](O)[C@@H](COP(O)(O)=O)OC1O