accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2(S)H2NOOHO

CAS RN 56-85-9

IUPAC Name: (2S)-2-amino-4-carbamoylbutanoic acid
Synonyms: L-glutamine glutamine express (genetic evolutionary nutrition) stimulina saforis glavamin 2-aminoglutaramic acid earthlink science glutamine chews chocolate (amerifit) levoglutamide Gln L-glutaminsaeure-5-amid
Molecular Weight (g/mol): 146.15
Molecular Formula: C5H10N2O3
InChi Key: ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES: N[C@@H](CCC(N)=O)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
  • (4)
  • (26)
Quantity 
  • (1)
  • (6)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (4)
Percent Purity 
  • (4)
  • (7)
  • (3)
  • (1)
  • (1)
  • (5)
Grade 
  • (8)
  • (3)
  • (1)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (11)
  • (6)
  • (4)
  • (6)
  • (3)
  • (2)
  • (5)
  • (2)
  • (4)
  • (26)
  • (1)
  • (6)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (4)
  • (4)
  • (7)
  • (3)
  • (1)
  • (1)
  • (5)
  • (8)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)