accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56-91-7

IUPAC Name: 4-(aminomethyl)benzoic acid
Synonyms: pamba 4-(aminomethyl)benzoesaeure styptopur amben gumbix α-amino- (8CI) p-toluic acid 氨甲苯酸 benzylamine-4-carboxylic acid aminomethylbenzoic acid p-toluic acid, α-amino- (8CI)
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: QCTBMLYLENLHLA-UHFFFAOYSA-N
SMILES: NCC1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results