accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HHClClNH2NHN

CAS RN 56-92-8

IUPAC Name: dihydrogen 2-(1H-imidazol-5-yl)ethan-1-amine dichloride
Synonyms: histamine dihydrochloride
Molecular Weight (g/mol): 184.06
Molecular Formula: C5H11Cl2N3
InChi Key: PPZMYIBUHIPZOS-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
Molecular Weight (g/mol) 
  • (6)
  • (7)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Certifications/Compliance 
  • (3)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (7)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)