accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 560-09-8

IUPAC Name: 1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid
Synonyms: camphoric acid
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: LSPHULWDVZXLIL-UHFFFAOYNA-N
SMILES: CC1(C)C(CCC1(C)C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results