accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSNH2

CAS RN 56004-83-2

IUPAC Name: 1-[2-(methylsulfanyl)phenyl]methanamine
Synonyms: 1-[2-(methylsulfanyl)phenyl]methanamine
Molecular Weight (g/mol): 153.24
Molecular Formula: C8H11NS
InChi Key: YVEJLBIEESKJLG-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1CN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results