accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56092-81-0

IUPAC Name: (4R,6S,8S,10Z,12R,14R,16E,18R,19R,20S,21S)-11,19,21-trihydroxy-22-[(2S,5'S)-5'-[(1R)-1-hydroxyethyl]-2,5'-dimethyl-[2,2'-bioxolan]-5-yl]-4,6,8,12,14,18,20-heptamethyl-9-oxodocosa-10,16-dienoic acid
Synonyms: ionomycin
Molecular Weight (g/mol): 709.02
Molecular Formula: C41H72O9
InChi Key: PGHMRUGBZOYCAA-OJFQOPKISA-N
SMILES: C[C@H](CCC(O)=O)C[C@H](C)C[C@H](C)C(=O)\C=C(/O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)CC1CC[C@](C)(O1)C1CC[C@](C)(O1)[C@@H](C)O
Quantity 
  • (2)
  • (1)
  • (2)
Product Type 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results