accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH2N

CAS RN 56129-55-6

IUPAC Name: 1-(pyridin-3-yl)ethan-1-amine
Synonyms: 1-(pyridin-3-yl)ethanamine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: IQVQNBXPYJGNEA-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results