accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56183-63-2

H3CCH3NH3CCH3PNCH3CH3H3CCH3Cl

CAS RN 56183-63-2

IUPAC Name: {[bis(propan-2-yl)amino](chloro)phosphanyl}bis(propan-2-yl)amine
Synonyms: [chloro(diisopropylamino)phosphanyl]diisopropylamine
Molecular Weight (g/mol): 266.79
Molecular Formula: C12H28ClN2P
InChi Key: FEHUTHGOLLQBNW-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)P(Cl)N(C(C)C)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results