accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONClOCl

CAS RN 56187-37-2

IUPAC Name: 2-(3,5-dichloro-4-oxo-1,4-dihydropyridin-1-yl)acetate
Synonyms: 2-(3,5-dichloro-4-oxopyridin-1-yl)acetate
Molecular Weight (g/mol): 221.01
Molecular Formula: C7H4Cl2NO3
InChi Key: MFMXEEQESUEMSB-UHFFFAOYSA-M
SMILES: [O-]C(=O)CN1C=C(Cl)C(=O)C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)