accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 562-54-9

IUPAC Name: potassium methyl sulfate
Synonyms: potassium methyl sulfate
Molecular Weight (g/mol): 150.19
Molecular Formula: CH3KO4S
InChi Key: WBGWGHYJIFOATF-UHFFFAOYSA-M
SMILES: [K+].COS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)