CAS RN 563-04-2
IUPAC Name:
tris(3-methylphenyl) phosphate
Synonyms:
tri-M-cresyl phosphate
Molecular Weight (g/mol):
368.37
Molecular Formula:
C21H21O4P
InChi Key:
RMLPZKRPSQVRAB-UHFFFAOYSA-N
SMILES:
CC1=CC(OP(=O)(OC2=CC(C)=CC=C2)OC2=CC=CC(C)=C2)=CC=C1
1
–
1
of
1
results
1
Phosphoric Acid Tris(3-methylphenyl) Ester, TRC
CAS: 563-04-2 Molecular Formula: C21 H21 O4 P Molecular Weight (g/mol): 368.36 Synonym: Phosphoric acid, tris(3-methylphenyl) ester (9CI, ACI),m-Tolyl phosphate, (C7H7O)3PO (6CI),Phosphoric acid, tri-m-tolyl ester (8CI),NSC 4055,TMCP,Tri-m-cresyl phosphate,Tri-m-tolyl phosphate,Tris(3-methylphenyl) phosphate,Tris(m-tolyl) phosphate,Tris-m-cresyl phosphate,Tri-3-cresyl phosphate IUPAC Name: tris(3-methylphenyl) phosphate SMILES: Cc1cccc(OP(=O)(Oc2cccc(C)c2)Oc3cccc(C)c3)c1