accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 563-12-2

IUPAC Name: O,O-diethyl [({[diethoxy(sulfanylidene)-λ⁵-phosphanyl]sulfanyl}methyl)sulfanyl]phosphonothioate
Synonyms: ethion diethion caswell no. 427 embathion fosfono 50 rodocid ethopaz ethodan rhodocide ethanox
Molecular Weight (g/mol): 384.46
Molecular Formula: C9H22O4P2S4
InChi Key: RIZMRRKBZQXFOY-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCSP(=S)(OCC)OCC
Molecular Weight (g/mol) 
  • (7)
  • (3)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (7)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)