accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOBr

CAS RN 563-70-2

IUPAC Name: bromo(nitro)methane
Synonyms: bromonitromethane
Molecular Weight (g/mol): 139.94
Molecular Formula: CH2BrNO2
InChi Key: DNPRVXJGNANVCZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)CBr
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results