accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNBr

CAS RN 56423-63-3

IUPAC Name: 2-bromopyrazine
Synonyms: 2-bromopyrazine
Molecular Weight (g/mol): 158.99
Molecular Formula: C4H3BrN2
InChi Key: WGFCNCNTGOFBBF-UHFFFAOYSA-N
SMILES: BrC1=NC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results