CAS RN 565-63-9
CAS RN 565-63-9
IUPAC Name:
(2Z)-2-methylbut-2-enoic acid
Synonyms:
angelic acid 2-methylisocrotonic acid angelicasaeure α-methyl isocrotonic acid acide angelique angelikasaeure cis-2-dimethylcrotonic acid acido angelico α-methylisocrotonic acid
Molecular Weight (g/mol):
100.12
Molecular Formula:
C5H8O2
InChi Key:
UIERETOOQGIECD-ARJAWSKDSA-N
SMILES:
C\C=C(\C)C(O)=O
1
–
1
of
1
results
1
Angelic Acid, TRC
CAS: 565-63-9 Molecular Formula: C5 H8 O2 Molecular Weight (g/mol): 100.12 Synonym: 2-Methylisocrotonic acid,cis-2,3-dimethylacrylic acid IUPAC Name: (Z)-2-methylbut-2-enoic acid SMILES: C\C=C(\C)/C(=O)O