accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 565-73-1

IUPAC Name: sodium carbamoylformate
Synonyms: sodium oxamate
Molecular Weight (g/mol): 111.03
Molecular Formula: C2H2NNaO3
InChi Key: RQVZIJIQDCGIKI-UHFFFAOYSA-M
SMILES: [Na+].NC(=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results