accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3COON(S)OHO

CAS RN 56502-01-3

IUPAC Name: 2-[(2S)-1-[(tert-butoxy)carbonyl]pyrrolidin-2-yl]acetic acid
Synonyms: [(2S)-1-(tert-butoxycarbonyl)pyrrolidin-2-yl]acetic acid
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: GDWKIRLZWQQMIE-QMMMGPOBSA-N
SMILES: CC(C)(C)OC(=O)N1CCC[C@H]1CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)