accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOO

CAS RN 5657-51-2

IUPAC Name: 5H,7H-furo[3,4-b]pyridin-5-one
Synonyms: 7H-furo[3,4-b]pyridin-5-one
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: YSJHADWSLVFGGT-UHFFFAOYSA-N
SMILES: O=C1OCC2=C1C=CC=N2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results