accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SCN

CAS RN 56601-42-4

IUPAC Name: isothiocyanatocyclopropane
Synonyms: isothiocyanatocyclopropane
Molecular Weight (g/mol): 99.15
Molecular Formula: C4H5NS
InChi Key: JGFBQFKZKSSODQ-UHFFFAOYSA-N
SMILES: S=C=NC1CC1
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)