accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56602-33-6

IUPAC Name: (1H-1,2,3-benzotriazol-1-yloxy)tris(dimethylamino)phosphanium; hexafluoro-λ⁵-phosphanuide
Synonyms: bop reagent
Molecular Weight (g/mol): 442.29
Molecular Formula: C12H22F6N6OP2
InChi Key: MGEVGECQZUIPSV-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CN(C)[P+](ON1N=NC2=CC=CC=C12)(N(C)C)N(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results