accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOO

CAS RN 5662-95-3

IUPAC Name: 8-oxaspiro[4.5]decane-7,9-dione
Synonyms: 8-oxaspiro[4.5]decane-7,9-dione
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: GFWLMILMVMCJDI-UHFFFAOYSA-N
SMILES: O=C1CC2(CCCC2)CC(=O)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results