accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56621-90-0

IUPAC Name: 2-chloropyrimidin-5-amine
Synonyms: 2-chloropyrimidin-5-amine
Molecular Weight (g/mol): 129.55
Molecular Formula: C4H4ClN3
InChi Key: DZBKIOJXVOECRA-UHFFFAOYSA-N
SMILES: NC1=CN=C(Cl)N=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results