accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 5663-96-7

IUPAC Name: oct-2-ynoic acid
Synonyms: 2-octynoic acid
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: BQDKCWCMDBMLEH-UHFFFAOYSA-N
SMILES: CCCCCC#CC(O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)