accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 56737-75-8

CH3NHNH2CH3HCl

CAS RN 56737-75-8

IUPAC Name: (2,3-dimethylphenyl)hydrazine hydrochloride
Synonyms: (2,3-dimethylphenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 172.66
Molecular Formula: C8H13ClN2
InChi Key: UKAYNIJBIXNDDJ-UHFFFAOYSA-N
SMILES: Cl.CC1=C(C)C(NN)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results