accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3NHH2N

CAS RN 56737-78-1

IUPAC Name: (2,5-dimethylphenyl)hydrazine
Synonyms: (2,5-dimethylphenyl)hydrazine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: XTXWNXNOTJBIIN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(NN)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results