CAS RN 56776-32-0
IUPAC Name:
hydrogen 6-chloro-N-ethyl-4-methyl-4-phenyl-4H-3,1-benzoxazin-2-amine chloride
Synonyms:
hydrogen etifoxine chloride
Molecular Weight (g/mol):
337.24
Molecular Formula:
C17H18Cl2N2O
InChi Key:
SCBJXEBIMVRTJE-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CCNC1=NC2=CC=C(Cl)C=C2C(C)(O1)C1=CC=CC=C1
1
–
2
of
2
results
1
Promotions Available
Tocris Bioscience™ Etifoxine hydrochloride
Tocris Bioscience™ Etifoxine hydrochloride
CAS: 56776-32-0 Molecular Formula: C17H18Cl2N2O
2
Etifoxine Hydrochloride, TRC
CAS: 56776-32-0 Molecular Formula: C17 H17 Cl N2 O . Cl H Molecular Weight (g/mol): 337.24 Synonym: 4H-3,1-Benzoxazin-2-amine, 6-chloro-N-ethyl-4-methyl-4-phenyl-, hydrochloride (1:1),4H-3,1-Benzoxazin-2-amine, 6-chloro-N-ethyl-4-methyl-4-phenyl-, monohydrochloride (9CI),Etifoxine hydrochloride,Stresam IUPAC Name: 6-chloro-N-ethyl-4-methyl-4-phenyl-3,1-benzoxazin-2-amine;hydrochloride SMILES: Cl.CCNC1=Nc2ccc(Cl)cc2C(C)(O1)c3ccccc3