CAS RN 56799-93-0
IUPAC Name:
N-(naphthalen-1-yl)-4-nitrobenzene-1-sulfonamide
Synonyms:
N-(naphthalen-1-yl)-4-nitrobenzenesulfonamide
Molecular Weight (g/mol):
328.34
Molecular Formula:
C16H12N2O4S
InChi Key:
PQKIMVCBRAJXGP-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC2=CC=CC=C12
1
–
1
of
1
results
1
N-(1-Naphthyl)-4-nitrobenzenesulfonamide, 97%, Thermo Scientific™
CAS: 56799-93-0 Molecular Formula: C16H12N2O4S Molecular Weight (g/mol): 328.342 MDL Number: MFCD01985189 InChI Key: PQKIMVCBRAJXGP-UHFFFAOYSA-N Synonym: n-naphthalen-1-yl-4-nitrobenzenesulfonamide,n-1-naphthyl-4-nitrobenzenesulfonamide,cbmicro_011238 PubChem CID: 879693 IUPAC Name: N-naphthalen-1-yl-4-nitrobenzenesulfonamide SMILES: C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=C(C=C3)[N+](=O)[O-]