CAS RN 568-72-9
IUPAC Name:
1,6,6-trimethyl-6H,7H,8H,9H,10H,11H-phenanthro[1,2-b]furan-10,11-dione
Synonyms:
tanshinone iia
Molecular Weight (g/mol):
294.35
Molecular Formula:
C19H18O3
InChi Key:
HYXITZLLTYIPOF-UHFFFAOYSA-N
SMILES:
CC1=COC2=C1C(=O)C(=O)C1=C3CCCC(C)(C)C3=CC=C21
1
–
3
of
3
results
1
Tanshinone IIA, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tanshinone IIA, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tanshinone IIA, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.