accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5680-80-8

IUPAC Name: methyl 2-amino-3-hydroxypropanoate hydrochloridyl
Synonyms: methyl 2-amino-3-hydroxypropanoate hydrochloridyl
Molecular Weight (g/mol): 154.57
Molecular Formula: C4H9ClNO3
InChi Key: QDXGLJWDLGMECL-UHFFFAOYNA-N
SMILES: [Cl].COC(=O)C(N)CO
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (3)