CAS RN 5683-31-8
IUPAC Name:
3-(trimethylsilyl)prop-2-ynoic acid
Synonyms:
3-(trimethylsilyl)prop-2-ynoic acid
Molecular Weight (g/mol):
142.23
Molecular Formula:
C6H10O2Si
InChi Key:
IPEATTYBFBRNEB-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C#CC(O)=O
1
–
1
of
1
results
1
3-(Trimethylsilyl)propiolic acid, 97%
CAS: 5683-31-8 Molecular Formula: C6H10O2Si Molecular Weight (g/mol): 142.23 MDL Number: MFCD00190216 InChI Key: IPEATTYBFBRNEB-UHFFFAOYSA-N Synonym: 3-trimethylsilyl propiolic acid,3-trimethylsilyl propynoic acid,trimethylsilylpropiolic acid,3-trimethylsilylpropynoic acid,3-trimethylsilyl prop-2-ynoic acid,2-propynoic acid, 3-trimethylsilyl,tms-propynoic acid,acmc-209lul,3-trimethylsilyl propynoicacid,3-trimethylsilyl propalic acid PubChem CID: 2760863 SMILES: C[Si](C)(C)C#CC(O)=O