accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5683-33-0

H3CNCH3N

CAS RN 5683-33-0

IUPAC Name: N,N-dimethylpyridin-2-amine
Synonyms: 2-dimethylaminopyridine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: PSHKMPUSSFXUIA-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC=N1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)