accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONNOO

CAS RN 5683-43-2

IUPAC Name: 2-methyl-6-nitro-1,3-benzoxazole
Synonyms: 2-methyl-6-nitro-1,3-benzoxazole
Molecular Weight (g/mol): 178.15
Molecular Formula: C8H6N2O3
InChi Key: DPVZLXWUFYMLBK-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(C=C2O1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results